VWR® Premium Mayo Hegar Needle Holder
Supplier: VWR International
10806-126 10807-434
10806-126EA 149.79 CAD
10806-126 10807-434
VWR® Premium Mayo Hegar Needle Holder
Forceps
VWR® Premium Mayo Hegar Needle Holders are constructed of surgical-grade German steel.
  • Passivated
  • Fully autoclavable
  • Box-lock joint
  • Rockwell hardness (HRC): 40–45
These reusable forceps can be sterilized by EtO/gamma radiation.