VWR® Mixter Forceps
Supplier: VWR International
10806-174 10806-176
10806-174EA 184.8 CAD
10806-174 10806-176
VWR® Mixter Forceps
Forceps
VWR® Mixter Forceps are constructed of surgical-grade stainless steel.
  • Passivated
  • Box-lock joint
  • Fully autoclavable
  • Full serrated jaws
  • Rockwell hardness (HRC): 40–45
These forceps can be sterilized by EtO/gamma radiation.