VWR® Rochester Carmalt Atraumatic Artery Forceps
Supplier: VWR International
10806-164 10806-302 10806-304 10806-306
10806-164EA 76.81 CAD
10806-164 10806-302 10806-304 10806-306
VWR® Rochester Carmalt Atraumatic Artery Forceps
Forceps
VWR® Rochester Carmalt Atraumatic Artery Forceps are constructed of surgical-grade stainless steel.
  • Passivated
  • Box-lock joint
  • Fully autoclavable
  • Surgical grade
  • Rockwell hardness (HRC): 40–45
These forceps can be sterilized by EtO/gamma radiation.