Polyethylene glycol 8,000 20% in 2.5 M sodium chloride, not sterile

Supplier: TEKNOVA CA

Synonyms: PEG 8000Ethylene glycol polymer 8000PEG 8,000

P4138 P4138 P4137 P4137
CA100216-878EA 352.53 CAD
100218-914 100216-878 CA100216-878 CA100218-914
Polyethylene glycol 8,000 20% in 2.5 M sodium chloride, not sterile
Polyethylene glycol 8,000

Polyethylene glycol (PEG) has a wide range of uses including cell fusion for the formation of hybridomas, precipitation of DNA, and to create macromolecular crowding in solutions.

Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR