Sklar Edge™ Tungsten Carbide Metzenbaum-Nelson Bariatric Scissors, OR Grade, Sklar
Supplier: Sklar
16-2210 16-2310 16-2311
11239-510EA 850.31 CAD
11239-510 11239-514 11239-516
Sklar Edge™ Tungsten Carbide Metzenbaum-Nelson Bariatric Scissors, OR Grade, Sklar
Scissors
Sklar Edge™ Tungsten Carbide Metzenbaum-Nelson Bariatric Scissors are long dissecting scissors most commonly used in Bariatric Surgeries, such as gastric bypass, gastric banding, and sleeve gastrectomy.
They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue. Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors which make them ideal for use in thoracic procedures as well. These scissors are not recommended for cutting sutures or drains. Sklar Edge™ TC Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision.
They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue
Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors which make them ideal for use in thoracic procedures as well. These scissors are not recommended for cutting sutures or drains. Sklar Edge™ TC Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision.