SpeedStreaks™ Disposable Loop
Supplier: HARDY DIAGNOSTICS CA
3153 179CSR40 8175CSR40H HS10F HS10R HS1F HS1R J87
CA10000-992EA 108.34 CAD
CA10000-992 CA10000-996 CA10000-998 CA10001-444 CA10001-446 CA10001-448 CA10001-450 CA10001-462
SpeedStreaks™ Disposable Loop
Spreaders Inoculating Loops and Needles
Hardy Diagnostic Loops are sterile and free of lubricants, oils, and electrostatic charges.
  • Loops and needle shaft improves grip
  • Packed in resealable plastic pouches
  • Non-calibrated inoculating loop used to transfer various liquids, suspensions, bacteria, and other specimens
  • Speedstreaks™ line of disposable inoculating loops
Hexagonal loop shaft improves grip, while the ultra smooth loop surfaces allow for easy streaking.
Hexagonal needle shaft also improves grip. Needles have a 1.45 mm diameter, 20 cm in length, are sterile and free of lubricants, oils, and electrostatic charges. Color coded for easy product identification. Packed in re-sealable plastic pouches for use with the LoopCaddy Stand.
Non-calibrated inoculating loop used to transfer various liquids, suspensions, bacteria, and other specimen from storage containers to culture media and/or petri dishes for purposes of study and measurement. Made from only high-grade nickel-chromium alloys to ensure repeated, non-calibrated usage.
SpeedStreaks™ are Hardy Diagnostics line of disposable inoculating loops. The loops are available in several styles and are color coded for convenience. The 1 and 10 µl loops, each come in two different rigidities, rigid-hard, or flexible-soft, to fit a variety of streaking preferences. An inoculating needle is also available formed in the rigid-hard plastic. The loops are manufactured using a process that smoothes the edges, free of plastic burrs and will not gouge the agar surface. The loops are free from coatings and electrostatic charges that might interfere with wetting and transfer of the volume contained in the loop. The 1ul and 10 µl loops are calibrated and each box of 1000 loops comes shipped with a Certificate of Calibration that authenticates the accuracy of the loops. The loops are certified to deliver a volume within 20% of their stated volume. The loops are certified using the Colorimetric Evans Blue Dye Procedure.