3510 White LD-100 Frock, Worklon®
Supplier: Superior Uniform
3510-S 3510-M 3510-L 3510-XL 3510-2XL 3510-3XL 3510-4XL 3510-5XL
89136-770EA 130.75 CAD
89136-770 89136-772 89136-774 89136-776 89136-778 89136-780 89136-782 89136-784
3510 White LD-100 Frock, Worklon®
Workwear
LD-100 polyester taffeta frocks are often used in pharmaceutical, medical device, food processing, and related industries where particulate control is required, and garments are donned in controlled environments up to and including Class 1000 (ISO 6)
The frocks feature a snap closure, raglan sleeves, polyester knit cuffs, two-ply reinforced placket, adjustable snaps at collar, and a color-coded hang loop denoting size.