MEINHARD™ Concentric Glass Nebulizer, Type C, PerkinElmer
Supplier: PerkinElmer
00472022 00472022
CA97012-890EA 1325.66 CAD
CA97012-890 97012-890
MEINHARD™ Concentric Glass Nebulizer, Type C, PerkinElmer
Atomizers
MEINHARD® Nebulizer made of glass for use with high dissolved solids solutions.
  • Simplicity of design and operation
  • Reproducible and self-aligning aerosol
  • Close fabrication control
Accessories information: Cleaner effectively used for MEINHARD®, MicroMist and SeaSpray nebulizers.