Sklarlite XD™ Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
23-1177 23-1178 23-1179
10195-102EA 430.83 CAD
10195-102 10195-104 10195-106
Sklarlite XD™ Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Scissors
Sklarlite XD™ Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue
These scissors are not recommended for cutting sutures, drains, or heavy tissue. Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors. Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today.