Surgi-OR™ TC Metzenbaum-Lahey Dissecting Scissors, Physician Grade, Sklar
Supplier: Sklar
95-119 95-120 95-122 95-335 95-336 95-339 95-340
89023-470EA 349.18 CAD
89023-470 89023-472 89023-474 89023-580 89023-582 89023-584 89023-586
Surgi-OR™ TC Metzenbaum-Lahey Dissecting Scissors, Physician Grade, Sklar
Scissors
Surgi-OR™ Metzenbaum-Lahey Dissecting Scissors are used for dissecting and cutting tissue only.
These scissors are not recommended for cutting sutures or drains. Metzenbaum-Lahey scissors tend to be shorter than standard Metzenbaum scissors. Hand-crafted in Pakistan, Surgi-OR™ instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards.