Sklar Black™ Potts-Smith Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
90-2304 90-2306
10199-110EA 685.46 CAD
10199-110 10199-112
Sklar Black™ Potts-Smith Dressing Forceps, OR Grade, Sklar
Forceps
Sklar Black™ Potts-Smith Dressing Forceps are most commonly used for application and changing of dressings in laser procedures
They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.