Disposable Tips for MLA and Ovation Pipettors, VistaLab
Supplier: VistaLab
4058-5000 4058-5332 4058-6000 4058-6100 4058-6102 4060-2004 4060-2132 4060-2332 4060-3004 4060-3132 4060-3332 4060-1333 4060-1002 4060-1032 4060-1332 4058-5102 4058-5133 4058-3000 4058-1000 4058-2000 4058-6133 4060-9024 4060-9025 4060-9026 2028 4026 4025 9048 2027 9025 9026 4070-1002LR 4070-1032LR 4070-2004LR 4070-2132LR 4070-3004LR 4070-3132LR 4070-1333LR 4070-1332LR 4070-2332LR 4070-3332LR 4058-6332 9050
89027-680PK 116.46 CAD
89027-680 89027-684 89049-042 89049-044 89049-046 89094-962 89094-964 89094-966 89094-970 89094-972 89094-974 89107-230 89107-232 89107-234 89107-236 97002-024 97002-026 14220-729 14220-735 14220-737 89049-048 89107-238 89107-240 89107-242 53514-114 53514-120 53514-110 53514-118 53514-124 53514-126 53514-144 53514-112 53514-154 53514-128 53514-130 53514-152 89368-676 89368-678 89368-680 89368-682 89368-684 89368-686 89368-688 89368-690 89368-692 89368-694 75813-808 75813-824
Disposable Tips for MLA and Ovation Pipettors, VistaLab
Pipette Tips
Disposable polypropylene tips for use with MLA or ovation pipettors.
  • Non-pyrogenic
  • Non-sterile or sterile, RNase/DNase-free
  • Filtered or non-filtered
  • Low retention available
  • Trace metal certified available
VistaLab tips are available in a full range of volumes from 2 μl to 10 ml for Macro Volume pipettors.
These disposable tips are an economical choice and are available in a wide range of sizes.
Order Now
SPECIFICATIONS
Environmentally Preferable