Ovation® Fixed Volume Pipettors, VistaLab
Supplier: VistaLab
1157-0005 1157-0010 1157-0020 1157-0025 1157-0100 1157-0200 1157-0250 1157-0300 1157-0500 1157-1000 1257-0001 1257-0002 1257-0005 1257-0010 1257-0020 1257-0025 1257-0100 1257-0200 1257-0250 1257-0300 1257-0500 1257-1000 1157-0999 1157-0050 1257-0050
14227-422EA 480.59 CAD
14227-422 14227-424 14227-426 14227-428 14227-432 14227-434 14227-436 14227-438 14227-440 14227-442 14227-444 14227-446 14227-448 14227-450 14227-452 14227-454 14227-458 14227-460 14227-462 14227-464 14227-466 14227-468 99089-398 75814-372 75814-376
Ovation® Fixed Volume Pipettors, VistaLab
Pipettes Repetitive Pipettors
Mechanical 1-stroke (F1) or 2-stroke (F2) pipettors with low-force plunger operation.
  • Ovation F1 models available in 11 fixed volumes ranging from 5 to 1000 μl
  • Ovation F2 models available in 13 fixed volumes ranging from 1 to 1000 μl
  • Built-in microtube cap opener
  • Custom fixed volumes available upon request
Ovation pipettors and essential components are available, with pipettors featuring mechanical low-pressure plunger operation.