MLA Macro Volume Pipettors, VistaLab
Supplier: VistaLab
1066 1068 1067 1061 1065 1069 1134 1079 1070 1076 1077 1078
53514-052EA 876.35 CAD
53514-052 53514-056 53514-054 53514-048 53514-050 53514-058 53514-078 75813-800 75814-394 75814-396 75814-398 75814-400
MLA Macro Volume Pipettors, VistaLab
Pipettes Repetitive Pipettors
MLA macro volume pipettors offer larger pipetting volumes than traditional models, with single-stroke 'to deliver' operation.
  • Available in a range of fixed and adjustable volumes
  • Different colors available for customizing models
MLA pipettors are made of precision-engineered aluminum alloy and stainless steel
Adjustable model available with a range from 1 to 5 ml in 0.1-ml increments.
Selectable model available with a range of set volumes at 2, 3, 4, and 5 ml.
MLA macro volume pipettors are available in 10 fixed volumes ranging from 1 to 10More Product Information