KIMAX® Erlenmeyer Flasks with Screw Caps, Kimcote®
Supplier: DWK Life Sciences (KIMBLE)
KC26720-250 KC26720-500 KC26720-1000 KC26720-2000
89097-392CS 686.09 CAD
89097-392 89097-394 89097-396 89097-398
KIMAX® Erlenmeyer Flasks with Screw Caps, Kimcote®
Flasks Erlenmeyer Flasks
Erlenmeyer flasks for preparing and storing media, and for use as shaker flasks.
  • KimCote® plastic safety coating help prevent and contain spills in broken glassware
  • Ultra-clear, extremely durable, and autoclavable
  • Large openings and GL 45 threaded closures for easy access and tight sealing
  • Marking spots and scales are durable white ceramic enamel
  • Made from 33 expansion borosilicate glass
Delivery information: Blue caps and clear pour rings included.