Racks for Upright Freezers, Chemglass
Supplier: Chemglass
CLS-3792-001 CLS-3792-003 CLS-3792-005 CLS-3792-007 CLS-3792-009 CLS-3792-011 CLS-3792-013 CLS-3792-015 CLS-3792-301
30623-554EA 230.68 CAD
30623-554 30623-556 30623-558 30623-560 30623-562 30623-564 30623-566 30623-568 30623-570
Racks for Upright Freezers, Chemglass
Racks Cryogenic Box Racks
Upright Freezer Drawer Racks are for standard 2" and 3" cardboard, plastic (polycarbonate) or metal (stainless steel or aluminum) boxes.
  • Ideal for −20 to −80 °C freezers
  • More organized system for storage and retrieval of samples
  • Constructed of corrosion resistant stainless steel, including handles and identification card holders
These racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer.