Addition Funnels with Glass Stopcock, Chemglass
Supplier: Chemglass
CG-1700-03 CG-1700-06 CG-1700-10 CG-1700-01 CG-1700-02 CG-1700-04 CG-1700-12 CG-1700-14 CG-1700-16
80070-478EA 637.87 CAD
80070-478 80070-484 80070-488 80070-474 80070-476 80070-480 80070-492 80070-496 80070-500
Addition Funnels with Glass Stopcock, Chemglass
Funnels Dropping Funnels
Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size.
  • With pressure equalizing arm and glass stopcock
Delivery information: Funnels with 14/20 and 19/22 joints are supplied complete with a stopper.