Density Sets

Supplier: Wards
PH0113F PH0113G PH0113H PH0113J
470221-106EA 109.65 CAD
470221-106 470221-108 470221-110 470221-112
Density Sets
Weights Calibration Weights
Students Learn That the Density of a Substance is Constant, Even as Its Mass and Volume Change.

  • Each set has 12 cylindrical bars
  • Bars are same diameter but different lengths
  • Each set comes in an attractive wooden holder
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR