Laboratory Jacks, Stainless Steel
Supplier: Wards
SSJCT10 SSJCT20 SSJCT30 SSJCT40 SSJCT50
470221-114EA 433.7 CAD
470221-114 470221-116 470221-118 470343-576 470345-402
Laboratory Jacks, Stainless Steel
Laboratory Jacks
Height-adjustable, corrosion-resistant stainless steel lab jacks capable of supporting heavy loads.
  • Adjustable lab jacks with screw-operated mechanism for easy, fluid movement of platform
  • Made of corrosion-resistant, easy-to-clean stainless steel
  • Gas springs to support load
  • Improved lift mechanism provides increased stability and torsional rigidity under load
  • 10 mm tapped hole for accepting support rod
  • Slip-proff rubber feet
  • Can be autoclaved
Strong, stable support for laboratory apparatus. The lab jack has a screw-operated scissor action mechanism for easy, fluid movement of platform - with and without load - and precise height adjustments. For physical, chemical, biological experiments and other uses.