Formaldehyde-Free Fetal Pigs
Supplier: VWR
WARD470000-442 WARD470001-720 WARD470001-716 140.1244.10 WARD470000-438 WARD470000-434 140.1235.1 WARD470001-724 WARD470001-718 WARD470000-458
470000-460PK 590.85 CAD
470000-460 470001-716 470001-724 470001-728 470001-718 470228-334 470000-458 470000-438 470228-332 470000-442 470001-720 470000-434
Formaldehyde-Free Fetal Pigs
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free preserved fetal pigs.
  • Mammalian anatomy
  • High quality preservation
To better serve the needs of all teaching environments and preferences, WARD’S offers this line as an alternative to our existing line of high-quality, safe preserved specimens. Absolutely no formaldehyde whatsoever is used in the fixing and preserving process for these specimens. Instead, our new solution used for preparation binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD'S offers the largest selection of specimens prepared with this alternative process, including the widest range of sizes and injection styles. In addition, the specimens can be used comfortably over extended periods of time.
Delivery information: Available in vacuum packs or pails.
Caution: Ward's preserved fetal pigs are a byproduct of the pork industry. Fetal pigs are not raised specifically for dissection. These animals are "recycled" for educational use.