Corning® Bar Coded External and Internal Threaded Polypropylene Cryogenic Vials, Corning
Supplier: Corning
8670 8671 8672 8676
75784-538CS 644.33 CAD
75784-538 75784-540 75784-542 75784-548
Corning® Bar Coded External and Internal Threaded Polypropylene Cryogenic Vials, Corning
Vials
These PP cryogenic vials with PP cap protect valuable cells, biological solutions, and aqueous solutions in ultra-low temperature storage.
  • 2 ml capacity, round bottom vials
  • Unique 1D and 2D barcodes, and human readable characters
  • Manufactured from polypropylene to withstand temperatures to as low as –196 °C (–321 °F)
  • Caps designed for a secure seal, and are compatible with automatic decapping equipment
  • Locking base
  • Sterile to SAL 10⁻⁶
  • Certified nonpyrogenic and DNase/RNase-free
The accessory cyrogenic vial boxes are designed for use with 1D and 2D barcoded vials. The 81-Place box accommodates internal or external threaded vials. The 100-Place box accommodates internal threaded vials. Boxes are autoclavable.