Polypropylene glycol, M.W. 2,000, Ultra Pure Grade
Supplier: VWR
Synonyms:
PPG
, Poly(propylene oxide)
VWRVE278-500G
10118-172EA
200.98
CAD
10118-172
Polypropylene glycol, M.W. 2,000, Ultra Pure Grade
Polypropylene glycol
Formula:
H(OCH₂CH₂CH₂)nOH Density: 1.005 g/cm³ (20 °C) Flash Pt: 112.8 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00084446 CAS Number: 25322-69-4 |
Specification Test Results
APHA |
|
Moisture (KF) |
|
Viscosity (@ 100° F) | 150 - 175 cS |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...