Polyvinyl alcohol, M.W. 500 - 5,000, high purity, hydrolysed

Supplier: VWR

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

9664-100G 9664-500G
10118-162CS 1110.03 CAD
10118-162 10118-164
Polyvinyl alcohol, M.W. 500 - 5,000, high purity, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Degree of Hydrolysis 85 - 89 %
Loss on Drying = 5.0 %
pH (4.0%, Water) @25C 4.5 - 7.0
Residue after Ignition = 2.0 %
Viscosity Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR