SP Wilmad-LabGlass Laboratory Grade Automatic Burettes, SP Industries
Supplier: SP Industries
LG-4240-102 LG-4240-104
14203-948EA 808.99 CAD
14203-948 14204-722 14207-294 14209-458 14203-736 14204-316
SP Wilmad-LabGlass Laboratory Grade Automatic Burettes, SP Industries
Burettes
This inexpensive portable titration set features a 2mm straight bore stopcock
The borosilicate glass burette is filled by pressure through a tube extending from the bottle up inside the graduated tube. A small hole in the filling tube at the "O" line permits setting of the liquid level and return of the excess to the bottle. Complete Set furnished with burette, bottle, rubber hose, rubber stopper, and rubber bulb.
Order Now
SPECIFICATIONS
Environmentally Preferable