SP Wilmad-LabGlass PTFE Beaded Fitting Couplings, SP Industries
Supplier: SP Industries
LG-4661-100 LG-4661-104
14200-782EA 261.35 CAD
14200-782 14200-820 14200-924 14200-946
SP Wilmad-LabGlass PTFE Beaded Fitting Couplings, SP Industries
Clamps Standard Taper and Spherical Joint Clamps
These PTFE/Viton® couplings provide an inert and greaseless connection for equipment with beaded fittings
Their unique design eliminates pressure points which cause glassware to break. Use with Wilmad-LabGlass beaded ends.