SP Wilmad-LabGlass Carboys with Hose Connection, SP Industries
Supplier: SP Industries
LG-3465-104 LG-3465-100
14205-194EA 1586.16 CAD
14205-194 14206-110 14207-556 14208-302
SP Wilmad-LabGlass Carboys with Hose Connection, SP Industries
Bottles Carboys
Large capacity borosilicate glass bottles, offered in four different sizes, feature a hose connector sealed at neck (¹/₂ or ³/₈" connection).
All bottle necks are compatible with a #12 stopper.