Budget Antibiotic Disks
Supplier: Ward's Science
XD1360-DISP XD1361-PK/50 XD1362-PK/50 XD1364-PK/50 XD1365-PK/50 XD1366-PK/50
470220-226EA 44.8 CAD
470220-226 470220-228 470220-230 470220-234 470220-236 470220-238
Budget Antibiotic Disks
Culture Dishes Petri Dishes
Users can observe the sensitivity of various bacteria and fungi to an array of antibiotics with these convenient kits.
  • Excellent Quality at an Economical Cost
  • Convenient and Reliable
  • Process Demonstrates Principles of Antibiotic Reactions
  • Fifty Reseable Disks in One Sleeve
Each resalable sleeve contains 50 discs of antibiotic; all you need to do is place the disc on the bacterial culture and observe the results to determine susceptibility or resistance of an organism to specific antibiotics.
Disc dispenser, required, is available separately.
Antibiotic Sensitivity Disc Set comes with 1 magazine of each of the following: Penicillin, Streptomycin, Tetracycline, Neomycin and Erythromycin