Anti-MRPL42 Mouse Monoclonal Antibody [clone: 3H6H2]

Supplier: GeneTex
GTX60760
10169-346EA 948.63 CAD
10169-346
Anti-MRPL42 Mouse Monoclonal Antibody [clone: 3H6H2]
Antibodies
Mouse Monoclonal Antibody [3H6H2] to MRPL42

Type: Primary
Antigen: MRPL42
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR