Anti-NR3C2 Mouse Monoclonal Antibody [clone: H10E4C9F]

Supplier: GeneTex
GTX22774
10168-492EA 997.75 CAD
10168-492
Anti-NR3C2 Mouse Monoclonal Antibody [clone: H10E4C9F]
Antibodies
Mouse monoclonal antibody [H10E4C9F] to Mineralocorticoid Receptor

Type: Primary
Antigen: NR3C2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Chicken, Rat, Rabbit, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR