Anti-TMED10 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX115517
10166-140EA 939.25 CAD
10166-140
Anti-TMED10 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TMP21 (transmembrane emp24-like trafficking protein 10 (yeast))

Type: Primary
Antigen: TMED10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR