Disposable Samplers, DispoPicker, Burkle
Supplier: BURKLE INC
5393-5540 5393-5541 5393-5550 5393-5551
10821-325PK 370.1 CAD
10821-325 10821-327 10821-329 10821-331
Disposable Samplers, DispoPicker, Burkle
Samplers Disposable Samplers
DispoPicker is ideally suited for cross-section samples.
  • HDPE
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated
Disposable sampler for powders and small-grain bulk materials. Because the tip is closed, the sample material cannot trickle back out of the sample lance.