Disposable Samplers, ViscoDispo, Burkle
Supplier: BURKLE INC
5393-3320 5393-3321 5393-3330 5393-3331
10821-303PK 499.93 CAD
10821-303 10821-305 10821-307 10821-339
Disposable Samplers, ViscoDispo, Burkle
Samplers Disposable Samplers
The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100.000 mPas.
  • HDPE
  • Without seals
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated
The ergonomically shaped handle and the very stable drawbar make it easy to draw even viscous substances.
Ordering information: Each Unit contains 20 packs.