Burets, Dr. Schilling Pattern, Schellbach, Burkle
Supplier: BURKLE INC
9695-3015 9695-3025 9695-3050
10810-026EA 257.84 CAD
10810-026 10810-038 10810-041
Burets, Dr. Schilling Pattern, Schellbach, Burkle
Burettes
Burets made of borosilicate glass 3.3 with splinter-proof sheathing.
  • Adjusted to Ex
  • Shatter-proof, resistant to chemicals
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
The titrating burets with splinter protection is a fast acting burette with Schellbach stripes based on Dr
The glass burette has a plastic sheathing which increases resistance to breaking and protects against splinters. The possibility of the burette breaking is much lower than with a conventional burette, and if it does break, there will be no dangerous splinters.
The Bürkle dosing armature allows for precise dosing and exact fine titration with the press button or micro-screw and precise discharge through the discharge jet.
Schilling’s design with automatic zero point extraction and Bürkle dosing armature.