MPure Tissue DNA Extraction Kit, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
117022400 117022400
10204-730EA 1019.45 CAD
10204-730 CA76046-198
MPure Tissue DNA Extraction Kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MPure Tissue DNA Extraction Kit is used with the MPure-12 nucleic acid purification system for the extraction and purification of genomic DNA from a variety of animal tissues, swab samples and blood stain.
  • Extraction of genomic DNA from animal tissues, swab samples, and blood stains
  • Completely automated purification protocol
  • Use with MPure-12 System
  • Run in as little as 40 minutes
  • Nucleic Acids can be used immediately in downstream applications
MPure™ Tissue DNA Extraction Kit contains everything needed for sample preparation. However, for tough tissues such as cardiac or skeletal muscle, spleen, intestinal or other connective tissue-rich samples, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time.
Capacity: 100-400 µL / 1 swab or brush for Dried Swab samples, 100-400 µL / 4 discs for dried blood, Purify DNA from 100-400 µL / 10-40mg of tissue
Sample Type: Animal Tissues, Blood Stain, Swab
Sample Volume: 100-400 µL
Packaging: Kit contains Lysis Buffer 2 = 720 µL, Binding Buffer 1 = 720 µL, Magnetic Bead Solution = 800 µL, Washing Buffer 1 = 1000 µL, Washing Buffer 2 = 1000 µL, Washing Buffer 3 = 1000 µL, Elution Buffer = 1000 µL, Elution Buffer = 1000 µL
Caution: Store at room temperature (15-25°C). Do not freeze the reagent cartridges. The Kits are stable for 12 months under this condition. Store the purified DNA at 4°C (short-term) or aliquot and store at -70°C (long-term) before performing downstream analysis