Blood DNA Extraction Kit 200, MPure™, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
MPurify Your Nucleic Acid: Obtain Superior Yield, Exceptional Purity, and Automated Performance in a Flexible, Convenient and Affordable Manner!
- Extraction of host gDNA, mitochondrial and viral DNA
- Extract DNA from 100-400µL mammalian whole blood
- Nucleic acids can be used immediately in downstream applications
- Use with MPure-12 System
- Completely automated
- Can be run un as little as 50 minutes
MPure Blood DNA Extraction Kit 200 is used with the MPure-12 aNAP System for extraction of DNA from 100-400 µL mammalian whole blood, suspension of mammalian blood cells. Nucleic acids extracted from MPure Blood DNA Extraction Kit 200 can be used in a number of downstream application including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, Southern Blot Analysis.
Content : Proteinase K solution = 40 µL, Lysis Buffer 2 = 1000 µL, Binding Buffer 1 = 600 µL, Magnetic Bead Solution = 800 µL, Washing Buffer 1 = 1000 µL, Washing Buffer 2 = 1000 µL, Washing Buffer 3 = 1000 µL, Elution Buffer 1 = 1000 µL, Elution Buffer 2 = 1000 µL
Caution: Store at room temperature (15-25°C). Do not freeze the reagent cartridges. The Kits are stable for 12 months under these conditions. Store the purified total nucleic acid at 4°C (short-term) or aliquot and store at –70°C (long-term) before performing the downstream analysis.