Lysing Matrix Z, MP Biomedicals
Supplier: MP BIOMEDICALS(FKA ICN BIO) CA
Lysing Matrix Z is used for breaking tough plant and animal samples.
- High density and high hardness
- Ideal for cracking very tough cell walls
- Smooth surface minimizes shearing
- Lysis of gram positive and gram negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence. Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.