Burpless® Ball Valve for Poly Drum Funnel, New Pig
Supplier: New Pig
DRM946
10860-618EA 449.56 CAD
10860-618
Burpless® Ball Valve for Poly Drum Funnel, New Pig
Valves Air Regulation Valves
Poly Ball Valve lets you open or close funnel in seconds with one hand.
  • For help meeting closed container regulations
  • PTFE seals
  • Open or close funnel in seconds
Add this valve to your Poly Burpless® Funnel for help meeting closed container regulations. Poly Ball Valve lets you open or close funnel in seconds with one hand. Screws into 2" NPT drum opening to prevent spills if drum falls over. Made of polyethylene with PTFE seals.