Steel Insect Pins
Supplier: United Scientific Supplies
IPIN00-PK/100 IPINS0-PK/100 IPIN01-PK/100 IPIN02-PK/100 IPIN03-PK/100 IPIN04-PK/100 IPIN05-PK/100 IPIN000-PK/100
470222-074PK 8.7 CAD
470222-074 470222-076 470222-078 470222-080 470222-082 470222-084 470222-086 470222-088
Steel Insect Pins
Specimen Mounting Supplies
These steel pins are coated with a corrosion-resistant finish and have a yellow epoxy head.
  • Corrosion-resistant finish
  • Sold as packs of 100
  • Yellow epoxy head
These steel pins are coated with a corrosion-resistant finish and have a yellow epoxy head. Sold as packs of 100.