Wobble-not Low-Insertion Force Serological Pipettes, VistaLab
Supplier: VistaLab
4090-0010 4090-0025 4090-0050 4090-0005 4290-0005 4290-0010 4290-0025 4290-0050 4190-0005 4190-0010 4190-0025 4190-0050 4090-0100 4190-0100 4290-0100
75794-572CS 249.09 CAD
75794-572 75794-574 75993-434 76176-750 76413-614 76413-616 76413-618 76413-620 76420-066 76420-068 76420-070 76420-072 76472-604 76472-606 76472-608
Wobble-not Low-Insertion Force Serological Pipettes, VistaLab
These pipettes feature a unique 2-tier step design at the plug end for less jamming and less dripping.
  • Sterile, RNase/DNase-free, Non-pyrogenic
  • Individually Wrapped in Paper/Plastic or Plastic/Plastic
  • Bulk packaging in plastic resealable bags
  • Filter at plug end made of porous polypropylene
  • Ultra-clear markings
Low-insertion force serological pipettes have an accuracy of ± 2% at full volume.