Poly IBC Spill Containment Pallet with Shelf, PIG®
Supplier: New Pig
PAK736-BK-WD PAK736-BK-WOD
10860-068EA 2843.05 CAD
10860-068 10860-070
Poly IBC Spill Containment Pallet with Shelf, PIG®
Spill Control Materials
This innovative design transfers weight to load bearing channels to maximize capacity.
  • Textured grating holds a variety of IBC sizes
  • Keep storage areas clean
  • Two-way forklift entry allows movement from either direction
Strong enough for long-term storage of IBCs, this innovative design transfers weight to load bearing channels to maximize capacity. Molded-in sump catches leaks, drips and spills to help you comply with regulations and keep storage areas clean. Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals. Textured grating holds a variety of IBC sizes; lifts out for easy access to sump. Low profile for easy access to and dispensing from IBC tanks. Two-way forklift entry allows movement from either direction when not loaded.