Dual Canister Sampling Manifold (Stainless Steel & Siltek®-Treated), Restek
Supplier: Restek
24999 24998
10862-924EA 553.35 CAD
10862-924 10862-940
Dual Canister Sampling Manifold (Stainless Steel & Siltek®-Treated), Restek
Manifolds
Field duplicates of canister samples frequently result in analyte concentrations with high relative standard deviations.
  • Duplicate sampling with all canister sizes using one or two flow controllers
  • Precise dimensions (9.5" wide x 3.5" high)
  • Provides accurate splitting of sample between two canisters
  • One-piece design means fewer leaks
  • Thick-walled stainless steel tubing is rugged enough for field use
In addition, field duplicates do not differentiate laboratory performance from sampling variability. Restek’s dual canister sampling manifold (DCSM)More Product Information