Cleaned Polyurethane Foam (PUF) Cartridges, Restek
Supplier: Restek
22117 22115 24295 22114 22116
10862-894EA 296.95 CAD
10862-894 10862-896 10862-904 10862-952 10863-242
Cleaned Polyurethane Foam (PUF) Cartridges, Restek
Samplers Air Samplers
These cartridges used for collection of semivolatiles (pesticides, PCBs, PAHs).
  • Precleaned and ready to use
  • Suitable for ambient, indoor, and industrial hygiene applications
  • PUF/XAD-2 "sandwiches" capture a wider range of semivolatiles
Both large high-volume (220 to 280 l/min) and small low-volume (1 to 5 l/min) PUFs available.