Canister Carrying Case, Restek
Supplier: Restek
24226
10862-808EA 994.74 CAD
10862-808
Canister Carrying Case, Restek
Carriers Transport Carriers
Fits two 6 L SilcoCan® or TO-Can® canisters tightly without foam.
  • Heavy-duty
  • All-aluminum design
No organic contaminants from foam or plastics.