Thermal Desorption Unit Tubes (Empty), Restek
Supplier: Restek
24055 24054
10862-732PK 1036.28 CAD
10862-732 10862-908
Thermal Desorption Unit Tubes (Empty), Restek
High-quality thermal desorption tubes are suitable for ppt to ppm concentrations of volatile organic compounds (VOCs) in ambient, indoor, personal, and industrial hygiene environments.
  • Use corrosion-resistant glass tubes for excellent inertness
  • Allow monitoring of sorbent bed condition
  • Choose stainless steel tubes for greater durability in the field
  • No sampling pump necessary for passive sampling with diffusion caps
  • Individually etched with unique serial number for convenient sample identification
  • The most reliable barcode "code 128" format on tube for recording and tracking
  • Each tube has an arrow indicating flow direction to reduce errors during use
  • Can be packed with any sorbent to suit any application
Fit Markes (ULTRA and UNITY™), PerkinElmer, and Shimadzu thermal desorbers
Empty tubes for direct desorption of VOCs from liquids, solids, or pastes. Available in stainless steel with front sorbent-retaining gauze, rear gauze, and gauze retaining spring supplied; or glass with glass frit positioned 15 mm from sampling end.
Packed tubes come with a report detailing the total mass of sorbent in the tube; conditioned tubes also include a blank chromatogram.
Certifications: US EPA TO-17; ASTM D6196; NIOSH 2549; DIN EN ISO 16017