Sample Inlets (Stainless Steel & Siltek® Treated), Restek
Supplier: Restek
26210 26209
10862-638EA 182.52 CAD
10862-638 10862-722 10862-750 10862-848
Sample Inlets (Stainless Steel & Siltek® Treated), Restek
Chromatography Supplies Chromatography Spare Parts
Sample inlets with fittings on each end.
  • Inlets have 1/4" stainless steel compression fitting on each end
  • One end connects to flow controller or canister
  • Nut on other end serves as rain guard
  • Includes nuts and ferrules
  • Two different lengths for use with large canisters and miniature canisters