Passive Air Sampling Kits-Soil Gas (Stainless Steel and Siltek®-Treated), Restek
Supplier: Restek
22931 22934 22933 22932 26336 22930 22936
10862-682KT 632.86 CAD
10862-682 10862-690 10862-692 10862-718 10862-768 10863-168 10863-196
Passive Air Sampling Kits-Soil Gas (Stainless Steel and Siltek®-Treated), Restek
Samplers Air Samplers
This unique grab sampler is specifically designed for soil gas sampling by allowing the connection of tubing coming from the soil gas sample port.
  • Stainless steel tee with orifice
  • Variety of orifice sizes for sampling from 4 minutes to 10 hours
  • 1 1/2" vacuum gauge (–30" Hg to 0" Hg)
The innovative design minimizes connections and leaks and houses a critical orifice in the tee fitting. It also incorporates a vacuum gauge and 2 µm frit filter. The 1/4" compression inlet and outlet allows easy connection to the canister valve and also to the tubing from the sample port. Several orifice sizes provide sampling times from 4 minutes to 10 hours on a 6 L canister. Individual replacement parts are available, providing a cost-effective alternative to replacing the entire sampler.
Accessories information: Air sampling canisters sold separately.