Veriflo® Flow Controllers (Stainless Steel and Siltek® Treated), Restek
Supplier: Restek
22102 24261 24255 22103 24263 24264 24262 24229 24239 24238 24260
10862-586EA 1760.3 CAD
10862-586 10862-700 10862-712 10862-992 10862-994 10862-996 10862-998 10863-036 10863-070 10863-192 10863-234
Veriflo® Flow Controllers (Stainless Steel and Siltek® Treated), Restek
Flow Controllers
Veriflo® 423XL flow controllers are offered in a Siltek®-treated and stainless steel version.
  • Treated and non-treated version
  • Available with or without critical orifice
The flow device is available with or without a critical orifice. The critical orifice in a Veriflo® flow controller is interchangeable.
Accessories information: Order orifices for alternate sampling times, or replacement orifices, separately.
Ordering information: Vacuum gauge sold separately.