Multi-Layer Foil Gas Sampling Bags, Restek
Supplier: Restek
22951 22950 22968 22966 22953 22967 22952
10862-576PK 196.09 CAD
10862-576 10862-578 10862-984 10862-986 10862-988 10862-990 10863-020
Multi-Layer Foil Gas Sampling Bags, Restek
Bags Sample Bags
Sampling bags are a low-cost, whole-air sampling device for high-level VOCs and permanent gases.
  • Good stability for low molecular weight compounds
  • Chemically inert with light and moisture protection
  • Not recommended for low ppm VOCs due to background levels
  • Protective five layer barrier minimizes gas permeability
  • 60 gauge nylon (outer layer)
  • Metalized aluminum/Polyethylene
  • 0.0003" aluminum foil/0.002" polyethylene (inner layer)
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions,More Product Information