Passive Air Sampling Kits-Grab (Stainless Steel and Siltek®-Treated), Restek
Supplier: Restek
26280 26282 26283 26285 26286 26287 26266 26263 26264 26265 26281 26267 26268 26269 26270 26284
10862-568EA 542.25 CAD
10862-568 10862-570 10862-572 10862-588 10862-590 10862-592 10862-594 10862-600 10862-602 10862-604 10862-912 10863-012 10863-014 10863-016 10863-018 10863-150
Passive Air Sampling Kits-Grab (Stainless Steel and Siltek®-Treated), Restek
Samplers Air Samplers
Passive air sampling kits for use with 1, 3, or 6 L canisters for grab air sampling.
  • Use with 1, 3, or 6 L canisters for grab air sampling
  • Variety of orifice sizes for fast sampling from 5 to 60 minutes
  • Connect 1/4" compression fitting directly to canister valve inlet
  • Replaceable frit protects orifice and valve from particulates
Accessories information: Air sampling canisters sold separately.