SilcoCan® Air Sampling Canisters with RAVE™ Valve, Restek
Supplier: Restek
22093 22090 22092 22091
10862-554EA 2530.89 CAD
10862-554 10862-564 10862-566 10862-724 10862-726 10862-734 10862-736 10862-756 10862-824 10862-826 10862-832 10862-834 10862-854 10862-860 10862-862 10862-864 10863-140 10863-142 10863-146 10863-188
SilcoCan® Air Sampling Canisters with RAVE™ Valve, Restek
Samplers Air Samplers
Ideal for low-level reactive sulfur (5–20 ppb), TO-14A, or TO-15 compounds.
  • For low-level compounds
  • Prevents sample adsorption
  • Excellent storage stability
  • High-quality seal
For ultimate inertness, SilcoCan® air sampling canisters feature our unique Siltek® treatment technology
Whether you are sampling for TO-14A, TO-15, or reactive sulfur compounds, SilcoCan® canisters are your best choice for inertness. In Tedlar® bags, the stability of low-level (100 ppbv) sulfur volatile organic compounds (VOCs) is poor, even within 24 hours of sampling. Sulfur compounds react with the metal surface in electropolished canisters, so they are unsuitable for collecting and storing low-level sulfur VOCs. SilcoCan® air sampling canisters, which feature a Siltek®-treated surface, offer excellent storage stability for sulfur VOCs at very low levels (5–20 ppbv), under dry or humid conditions. The versatility of the SilcoCan® canister makes it an excellent choice for collecting and storing TO-14A or TO-15 compounds.
Canisters and valves made of 304 and 316 stainless steel to withstand the rigors of field work. Both 2-port and 3-port valves available; 3-port valve includes -30" Hg/60 psi vacuum/pressure gauge (other gauges available).